Product Details

SNP ID
rs61742046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77407557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACGCAGATATTAGGCAGCTGAGA[C/T]GGCAAGCAGCAGAGGAATATCACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOXRED1 PubMed Links

Gene Details

Gene
NOXRED1
Gene Name
NADP dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113475.2 518 Silent Mutation CCA,CCG P146P NP_001106946.1
XM_005267330.3 518 Silent Mutation CCA,CCG P146P XP_005267387.1
XM_011536427.2 518 Silent Mutation CCA,CCG P146P XP_011534729.1
XM_011536428.2 518 Silent Mutation CCA,CCG P146P XP_011534730.1
XM_011536429.2 518 Silent Mutation CCA,CCG P56P XP_011534731.1
XM_017020969.1 518 Silent Mutation CCA,CCG P146P XP_016876458.1
XM_017020970.1 518 Intron XP_016876459.1

View Full Product Details