Product Details
- SNP ID
-
rs4073612
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:94132485 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGACGTGTGAGACAACATGTGGCT[G/T]CTGGAGCTTTAGTGGCACCGTCACA
- Phenotype
-
MIM: 608420
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PANX1
PubMed Links
Gene Details
- Gene
- PANX1
- Gene Name
- pannexin 1
View Full Product Details