Product Details

SNP ID
rs12948434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42123168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAAGGTGCACCGGAAAATGCT[A/C]CCGTCCAACCTGAGTCCTACCGCCA
Phenotype
MIM: 608344 MIM: 602301 MIM: 604037
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSPB9 PubMed Links

Gene Details

Gene
HSPB9
Gene Name
heat shock protein family B (small) member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033194.2 431 Silent Mutation CTA,CTC L106L NP_149971.1
Gene
KAT2A
Gene Name
lysine acetyltransferase 2A
There are no transcripts associated with this gene.

Gene
RAB5C
Gene Name
RAB5C, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details