Product Details
- SNP ID
-
rs11992923
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:142212197 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCTCTCGGGTCCATTGCAGGCCA[A/G]GTGACGGCATCAGCTGACAACGCAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00051
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78564166] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00051
- Gene Name
- long intergenic non-protein coding RNA 51
There are no transcripts associated with this gene.
- Gene
- TSNARE1
- Gene Name
- t-SNARE domain containing 1
View Full Product Details