Product Details

SNP ID
rs3923786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136250450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCGCTGCAAATCTGCGGGGCTTC[A/G]GTGGTGGAGATTTATCGTCATGCCA
Phenotype
MIM: 612860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESP33 PubMed Links

Gene Details

Gene
ESP33
Gene Name
uncharacterized locus ESP33
There are no transcripts associated with this gene.

Gene
LOC101928722
Gene Name
nascent polypeptide-associated complex subunit alpha, muscle-specific form-like
There are no transcripts associated with this gene.

Gene
QSOX2
Gene Name
quiescin sulfhydryl oxidase 2
There are no transcripts associated with this gene.

View Full Product Details