Product Details

SNP ID
rs36101985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142205438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGGGGTCTCCACTTGGTGGTG[C/T]TGGGGGCCCCACCTGAGCCCTGGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00051 PubMed Links

Gene Details

Gene
LINC00051
Gene Name
long intergenic non-protein coding RNA 51
There are no transcripts associated with this gene.

Gene
TSNARE1
Gene Name
t-SNARE domain containing 1
There are no transcripts associated with this gene.

View Full Product Details