Product Details
- SNP ID
-
rs6527730
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:16807752 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCCTCTGAAGGTCCACTCTAGCT[A/C]TTGCTGGCAAGGACTGGGTAATTTG
- Phenotype
-
MIM: 300677
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TXLNG
PubMed Links
Gene Details
- Gene
- TXLNG
- Gene Name
- taxilin gamma
View Full Product Details