Product Details

SNP ID
rs13077644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113748930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGATCTCTAGAGGAGAGGGGAA[A/G]AAAAAAAGGAAGTGTTACTTCAGTG
Phenotype
MIM: 607027 MIM: 610834
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1A PubMed Links

Gene Details

Gene
ATP6V1A
Gene Name
ATPase H+ transporting V1 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001690.3 Intron NP_001681.2
Gene
NAA50
Gene Name
N(alpha)-acetyltransferase 50, NatE catalytic subunit
There are no transcripts associated with this gene.

View Full Product Details