Product Details

SNP ID
rs4499339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40299414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAACCAGACTAATTATTAGTGAC[C/G]TAAATTAATAGTTACCTGAGATCCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf47 PubMed Links

Gene Details

Gene
C19orf47
Gene Name
chromosome 19 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256440.1 Intron NP_001243369.1
NM_001256441.1 Intron NP_001243370.1

View Full Product Details