Product Details

SNP ID
rs1133442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:19980023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGAGTTTAATGGTACTTCATTC[A/G]AAATGGGAAAGAACTTGCCTTTTCT
Phenotype
MIM: 179512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PP2D1 PubMed Links

Gene Details

Gene
PP2D1
Gene Name
protein phosphatase 2C like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252657.1 Intron NP_001239586.1
Gene
RAB5A
Gene Name
RAB5A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292048.1 Intron NP_001278977.1
NM_004162.4 Intron NP_004153.2

View Full Product Details