Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001297696.1 | 2995 | UTR 3 | NP_001284625.1 | ||
NM_001297697.1 | 2995 | UTR 3 | NP_001284626.1 | ||
NM_176816.4 | 2995 | UTR 3 | NP_789786.2 | ||
XM_005248458.4 | 2995 | UTR 3 | XP_005248515.1 | ||
XM_005248459.4 | 2995 | UTR 3 | XP_005248516.1 | ||
XM_005248461.3 | 2995 | UTR 3 | XP_005248518.1 | ||
XM_005248463.3 | 2995 | UTR 3 | XP_005248520.1 | ||
XM_005248464.3 | 2995 | Intron | XP_005248521.1 | ||
XM_006714569.3 | 2995 | UTR 3 | XP_006714632.1 | ||
XM_006714570.3 | 2995 | UTR 3 | XP_006714633.1 | ||
XM_011543255.2 | 2995 | UTR 3 | XP_011541557.1 | ||
XM_011543256.2 | 2995 | UTR 3 | XP_011541558.1 | ||
XM_011543257.2 | 2995 | UTR 3 | XP_011541559.1 | ||
XM_011543258.2 | 2995 | Intron | XP_011541560.1 | ||
XM_011543259.2 | 2995 | Intron | XP_011541561.1 | ||
XM_011543260.2 | 2995 | Intron | XP_011541562.1 | ||
XM_017009206.1 | 2995 | UTR 3 | XP_016864695.1 | ||
XM_017009207.1 | 2995 | Intron | XP_016864696.1 | ||
XM_017009208.1 | 2995 | UTR 3 | XP_016864697.1 | ||
XM_017009209.1 | 2995 | UTR 3 | XP_016864698.1 | ||
XM_017009210.1 | 2995 | UTR 3 | XP_016864699.1 | ||
XM_017009211.1 | 2995 | Intron | XP_016864700.1 |