Product Details

SNP ID
hCV26228459
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.1:207508156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTTAGGGAAGAAGGGGATGAAT[C/G]GGAAGAGCACAGAGGATTTTGAGGG
Phenotype
MIM: 120620
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CR1 PubMed Links

Gene Details

Gene
CR1
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
There are no transcripts associated with this gene.

View Full Product Details