Product Details

SNP ID
hCV26241326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:18987874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAATGAAAGAACCTGGAAGGGCC[C/T]TGATATGACTGTGGTTGGACTGACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMC7 PubMed Links

Gene Details

Gene
TMC7
Gene Name
transmembrane channel like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160364.2 Intron NP_001153836.1
NM_001300732.1 Intron NP_001287661.1
NM_001324263.1 Intron NP_001311192.1
NM_001324265.1 Intron NP_001311194.1
NM_001324268.1 Intron NP_001311197.1
NM_024847.4 Intron NP_079123.3

View Full Product Details