Product Details

SNP ID
rs2635548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19821874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATAGAGAGATTTTAAAATCTAAG[A/G]TTCCTCATGTGGCAATGAACATTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4N2 PubMed Links

Gene Details

Gene
OR4N2
Gene Name
olfactory receptor family 4 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004723.2 Intron NP_001004723.1

View Full Product Details