Product Details

SNP ID
rs11864709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21234522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACACACACACACACACACACACAC[C/G]CCATCTCTTTGCCCAGATTTTAATG
Phenotype
MIM: 609901
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKS4B PubMed Links

Gene Details

Gene
ANKS4B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145865.2 Intron NP_665872.2

View Full Product Details