Product Details

SNP ID
rs2509979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62437141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACAGAATCATGATTTGACTCAT[C/T]ATAGATCAGCTTTGCCTCTTTCTGA
Phenotype
MIM: 103390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHNAK PubMed Links

Gene Details

Gene
AHNAK
Gene Name
AHNAK nucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001620.2 Intron NP_001611.1
NM_024060.3 Intron NP_076965.2
XM_005274240.1 Intron XP_005274297.1
XM_005274241.2 Intron XP_005274298.1
XM_005274245.1 Intron XP_005274302.1
XM_017018270.1 Intron XP_016873759.1
XM_017018271.1 Intron XP_016873760.1
XM_017018272.1 Intron XP_016873761.1
XM_017018273.1 Intron XP_016873762.1

View Full Product Details