Product Details

SNP ID
rs1047546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:79590134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGCTATCATTAAAGGCCAAAAAA[A/T]TCACTGCTAGAAATGTTCCCCAAAA
Phenotype
MIM: 601936
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PAWR PubMed Links

Gene Details

Gene
PAWR
Gene Name
pro-apoptotic WT1 regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002583.2 3662 Intron NP_002574.2
XM_006719435.2 3662 UTR 3 XP_006719498.1
XM_006719436.3 3662 Intron XP_006719499.1
XM_017019377.1 3662 UTR 3 XP_016874866.1
XM_017019378.1 3662 Intron XP_016874867.1
XM_017019379.1 3662 Intron XP_016874868.1

View Full Product Details