Product Details

SNP ID
rs12820313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95861926 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGCCATCACCGTTGTCCATTTC[C/T]GGAACTTGTTTACCATCCAAAAGTG
Phenotype
MIM: 603541
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC38 PubMed Links

Gene Details

Gene
CCDC38
Gene Name
coiled-coil domain containing 38
There are no transcripts associated with this gene.

Gene
LOC105369921
Gene Name
uncharacterized LOC105369921
There are no transcripts associated with this gene.

Gene
SNRPF
Gene Name
small nuclear ribonucleoprotein polypeptide F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003095.2 Intron NP_003086.1

View Full Product Details