Product Details

SNP ID
rs2272533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70293488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCGCGGGAGCCTGGCCGGGATC[C/T]CGGCGACCTCGGACCGGAAGAAGAA
Phenotype
MIM: 616275 MIM: 603542
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM136A PubMed Links

Gene Details

Gene
FAM136A
Gene Name
family with sequence similarity 136 member A
There are no transcripts associated with this gene.

Gene
SNRPG
Gene Name
small nuclear ribonucleoprotein polypeptide G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317165.1 Intron NP_001304094.1
NM_001317166.1 Intron NP_001304095.1
NM_001317167.1 Intron NP_001304096.1
NM_001317168.1 Intron NP_001304097.1
NM_001317169.1 Intron NP_001304098.1
NM_001317171.1 Intron NP_001304100.1
NM_003096.3 Intron NP_003087.1
XM_017004774.1 Intron XP_016860263.1

View Full Product Details