Product Details

SNP ID
rs12807306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94130647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCATCGGCTGAGGTCTTGTGCA[C/T]AGAGGGTGTGAAGAGTGGAAGAAAA
Phenotype
MIM: 608420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PANX1 PubMed Links

Gene Details

Gene
PANX1
Gene Name
pannexin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015368.3 Intron NP_056183.2
XM_011542734.2 Intron XP_011541036.1
XM_017017464.1 Intron XP_016872953.1

View Full Product Details