Product Details

SNP ID
rs2101313
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:207088949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTCTGGAGGGACAGAGACAGGT[A/G]AGGTTAGTTTACTTCCTTGAGCTTT
Phenotype
MIM: 120831 MIM: 171835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4BPB PubMed Links

Gene Details

Gene
C4BPB
Gene Name
complement component 4 binding protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000716.3 Intron NP_000707.1
NM_001017364.1 Intron NP_001017364.1
NM_001017365.2 Intron NP_001017365.1
NM_001017366.2 Intron NP_001017366.1
NM_001017367.1 Intron NP_001017367.1
XM_005273254.4 Intron XP_005273311.1
XM_005273255.2 Intron XP_005273312.1
Gene
PFKFB2
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2
There are no transcripts associated with this gene.

View Full Product Details