Product Details

SNP ID
rs2707846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66731427 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAACGCCTGGCTGGTAAATAAGG[A/T]TCTTGGAGGCTGTGCATGATGGCTC
Phenotype
MIM: 609700
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RABGEF1 PubMed Links

Gene Details

Gene
RABGEF1
Gene Name
RAB guanine nucleotide exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287060.1 Intron NP_001273989.1
NM_001287061.1 Intron NP_001273990.1
NM_001287062.1 Intron NP_001273991.1
NM_014504.2 Intron NP_055319.1

View Full Product Details