Product Details

SNP ID
rs1811845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54925994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGGAGTGCCTGGCCTGGCTTTG[A/G]CAGAGTGGTTCTCAAGGAGGGCTTC
Phenotype
MIM: 311790 MIM: 300132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PFKFB1 PubMed Links

Gene Details

Gene
PFKFB1
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
There are no transcripts associated with this gene.

Gene
TRO
Gene Name
trophinin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039705.2 Intron NP_001034794.1
NM_001271183.1 Intron NP_001258112.1
NM_001271184.1 Intron NP_001258113.1
NM_016157.3 Intron NP_057241.2
NM_177556.2 Intron NP_808224.1
NM_177557.2 Intron NP_808225.1
XM_006724600.2 Intron XP_006724663.1
XM_011530808.1 Intron XP_011529110.1
XM_011530809.1 Intron XP_011529111.1
XM_011530811.2 Intron XP_011529113.1
XM_011530812.1 Intron XP_011529114.1
XM_011530813.1 Intron XP_011529115.1
XM_011530814.1 Intron XP_011529116.1
XM_017029767.1 Intron XP_016885256.1
XM_017029768.1 Intron XP_016885257.1
XM_017029769.1 Intron XP_016885258.1
XM_017029770.1 Intron XP_016885259.1
XM_017029771.1 Intron XP_016885260.1
XM_017029772.1 Intron XP_016885261.1
XM_017029773.1 Intron XP_016885262.1

View Full Product Details