Product Details

SNP ID
rs3820321
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:20636334 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGCTTCTGCTCCATGAAGTTTATAA[A/G]GGAAGTTGGATTTTCCTTTTTTTTT
Phenotype
MIM: 608309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6084 PubMed Links

Gene Details

Gene
MIR6084
Gene Name
microRNA 6084
There are no transcripts associated with this gene.

Gene
PINK1
Gene Name
PTEN induced putative kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032409.2 Intron NP_115785.1
Gene
PINK1-AS
Gene Name
PINK1 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details