Product Details

SNP ID
rs4574706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:72746750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACTAGAGCCCATAAGATGGAAG[A/T]GCAACCAAGGTAGCTTCGCGAAAGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TYW1B PubMed Links

Gene Details

Gene
TYW1B
Gene Name
tRNA-yW synthesizing protein 1 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145440.2 Intron NP_001138912.2
XM_011516230.2 Intron XP_011514532.1
XM_011516231.2 Intron XP_011514533.1
XM_011516232.2 Intron XP_011514534.1
XM_011516233.2 Intron XP_011514535.1
XM_017012221.1 Intron XP_016867710.1
XM_017012222.1 Intron XP_016867711.1

View Full Product Details