Product Details

SNP ID
rs1015011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24497053 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTCTTCCTCACCTTCTCTCCCC[C/G]CTATCTCCCTTCTGTTTTTCAACGG
Phenotype
MIM: 606673
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADORA2A-AS1 PubMed Links

Gene Details

Gene
ADORA2A-AS1
Gene Name
ADORA2A antisense RNA 1
There are no transcripts associated with this gene.

Gene
UPB1
Gene Name
beta-ureidopropionase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016327.2 Intron NP_057411.1
XM_011530222.2 Intron XP_011528524.1
XM_011530223.2 Intron XP_011528525.1
XM_011530224.2 Intron XP_011528526.1
XM_011530225.2 Intron XP_011528527.1
XM_017028825.1 Intron XP_016884314.1
XM_017028826.1 Intron XP_016884315.1
XM_017028827.1 Intron XP_016884316.1
XM_017028828.1 Intron XP_016884317.1

View Full Product Details