Product Details
- SNP ID
-
rs28364866
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:681193 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGGGTGGAATGGAGAGGGGGGCT[T/C]ACGGGGGGCGGGGGGGTGCTTGTGT
- Phenotype
-
MIM: 605343
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FSTL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28364867] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSTL3
- Gene Name
- follistatin like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005860.2 |
|
Intron |
|
|
NP_005851.1 |
- Gene
- PRSS57
- Gene Name
- protease, serine 57
There are no transcripts associated with this gene.
View Full Product Details