Product Details

SNP ID
rs7989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95237639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACACGTTATATTTGATTGGGTT[A/C]AAATTTGGCTGATGTCCAAATGCAG
Phenotype
MIM: 605900
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACSM6 PubMed Links

Gene Details

Gene
ACSM6
Gene Name
acyl-CoA synthetase medium-chain family member 6
There are no transcripts associated with this gene.

Gene
PDLIM1
Gene Name
PDZ and LIM domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020992.3 1215 Intron NP_066272.1
XM_011540330.1 1215 UTR 3 XP_011538632.1

View Full Product Details