Product Details

SNP ID
rs2780159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83860868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTCTCTAGTAGTTTCCCTGGA[C/T]AGGCTTGATGGGAGATTTTTTTTTT
Phenotype
MIM: 611253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF27 PubMed Links

Gene Details

Gene
KIF27
Gene Name
kinesin family member 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271927.1 Intron NP_001258856.1
NM_001271928.1 Intron NP_001258857.1
NM_017576.2 Intron NP_060046.1
XM_011518848.2 Intron XP_011517150.1
XM_011518849.2 Intron XP_011517151.1
XM_011518850.2 Intron XP_011517152.1
XM_011518854.2 Intron XP_011517156.1
XM_011518856.2 Intron XP_011517158.1
XM_011518857.2 Intron XP_011517159.1
XM_011518858.2 Intron XP_011517160.1
XM_017014900.1 Intron XP_016870389.1
XM_017014901.1 Intron XP_016870390.1
XM_017014902.1 Intron XP_016870391.1
XM_017014903.1 Intron XP_016870392.1
XM_017014904.1 Intron XP_016870393.1
XM_017014905.1 Intron XP_016870394.1
XM_017014906.1 Intron XP_016870395.1
XM_017014907.1 Intron XP_016870396.1
XM_017014908.1 Intron XP_016870397.1
XM_017014909.1 Intron XP_016870398.1
XM_017014910.1 Intron XP_016870399.1
XM_017014911.1 Intron XP_016870400.1
XM_017014912.1 Intron XP_016870401.1
XM_017014913.1 Intron XP_016870402.1
XM_017014914.1 Intron XP_016870403.1
XM_017014915.1 Intron XP_016870404.1
XM_017014916.1 Intron XP_016870405.1

View Full Product Details