Product Details

SNP ID
rs2634204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6449991 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAATCCTGTGACTCCCCATAGCTC[C/T]TGGGAGGAGAGCAAGCCCCTTAAGG
Phenotype
MIM: 142290 MIM: 605493
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPX PubMed Links

Gene Details

Gene
HPX
Gene Name
hemopexin
There are no transcripts associated with this gene.

Gene
TRIM3
Gene Name
tripartite motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248006.1 Intron NP_001234935.1
NM_001248007.1 Intron NP_001234936.1
NM_006458.3 Intron NP_006449.2
NM_033278.3 Intron NP_150594.2
XM_011519844.1 Intron XP_011518146.1
XM_011519845.2 Intron XP_011518147.1
XM_017017098.1 Intron XP_016872587.1
XM_017017099.1 Intron XP_016872588.1

View Full Product Details