Product Details

SNP ID
hCV26928088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:67678217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCAAGGCTGGAAGGTTTTGCT[C/T]TCTTTGTGCTAAAGGTTTTGAAAAC
Phenotype
MIM: 603930 MIM: 607849 MIM: 603207
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPHN PubMed Links

Gene Details

Gene
GPHN
Gene Name
gephyrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024218.1 1151 Intron NP_001019389.1
NM_020806.4 1151 Intron NP_065857.1
Gene
RDH11
Gene Name
retinol dehydrogenase 11 (all-trans/9-cis/11-cis)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252650.1 1151 UTR 3 NP_001239579.1
NM_016026.3 1151 UTR 3 NP_057110.3
Gene
VTI1B
Gene Name
vesicle transport through interaction with t-SNAREs 1B
There are no transcripts associated with this gene.

View Full Product Details