Product Details

SNP ID
rs2013452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144003245 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTAGATAGATGAAGGGTTTGAA[A/G]CAGTTTTTTATTTAGGGAAAATCAC
Phenotype
MIM: 111740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GYPB PubMed Links

Gene Details

Gene
GYPB
Gene Name
glycophorin B (MNS blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304382.1 Intron NP_001291311.1
NM_002100.5 Intron NP_002091.3
XM_011531903.2 Intron XP_011530205.1
XM_011531904.2 Intron XP_011530206.1
XM_011531905.2 Intron XP_011530207.1
XM_017008137.1 Intron XP_016863626.1

View Full Product Details