Product Details

SNP ID
rs7676146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68821257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAATTTATATATTGGCATACAT[C/T]GGGGAACTCAAGTCAGAATAATTCT
Phenotype
MIM: 600070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGT2B10 PubMed Links

Gene Details

Gene
UGT2B10
Gene Name
UDP glucuronosyltransferase family 2 member B10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001075.5 Intron NP_001066.1
NM_001144767.2 Intron NP_001138239.1
NM_001290091.1 Intron NP_001277020.1
XM_017008585.1 Intron XP_016864074.1

View Full Product Details