Product Details

SNP ID
rs4130904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111388275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCAGTAGATCTGGAGTGGCCAT[G/T]ACTCAGTTGGTTGGTTGGTTGGTTG
Phenotype
MIM: 605673 MIM: 601206
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BTG4 PubMed Links

Gene Details

Gene
BTG4
Gene Name
BTG anti-proliferation factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017589.3 Intron NP_060059.1
XM_011542876.2 Intron XP_011541178.1
XM_011542878.2 Intron XP_011541180.1
XM_011542879.2 Intron XP_011541181.1
XM_011542880.2 Intron XP_011541182.1
Gene
POU2AF1
Gene Name
POU class 2 associating factor 1
There are no transcripts associated with this gene.

View Full Product Details