Product Details
- SNP ID
-
rs4407204
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:237086165 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGGAGCTCACGGGGCGGGCGTGG[A/G]AGGTGGACGTGTGGATCCCTGACCG
- Phenotype
-
MIM: 616011
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COPS8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148065526] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COPS8
- Gene Name
- COP9 signalosome subunit 8
View Full Product Details