Product Details
- SNP ID
-
rs6059892
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:34517961 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCACATTTTCTTTATCCATTTACCT[A/G]TTGATGGACGCTAATTCCATAATTT
- Phenotype
-
MIM: 607167
MIM: 606409
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DYNLRB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73259114] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DYNLRB1
- Gene Name
- dynein light chain roadblock-type 1
- Gene
- ITCH
- Gene Name
- itchy E3 ubiquitin protein ligase
There are no transcripts associated with this gene.
View Full Product Details