Product Details

SNP ID
rs12901354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52111507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCATTGAATCAACGCCAATAAG[A/G]TGGTGGGGAAAGAAATGGGGTCTAG
Phenotype
MIM: 606910 MIM: 604447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L10 PubMed Links

Gene Details

Gene
BCL2L10
Gene Name
BCL2 like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306168.1 Intron NP_001293097.1
NM_020396.3 Intron NP_065129.1
Gene
GNB5
Gene Name
G protein subunit beta 5
There are no transcripts associated with this gene.

View Full Product Details