Product Details

SNP ID
rs2114572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:58690875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGTAGATGCAGCTGGATAGGT[C/T]TGAGGATCCTCAAGGGAGAAGCCAG
Phenotype
MIM: 602192
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM10 PubMed Links

Gene Details

Gene
ADAM10
Gene Name
ADAM metallopeptidase domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110.3 Intron NP_001101.1
NM_001320570.1 Intron NP_001307499.1
Gene
HSP90AB4P
Gene Name
heat shock protein 90 alpha family class B member 4, pseudogene
There are no transcripts associated with this gene.

View Full Product Details