Product Details
- SNP ID
-
rs6461807
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:902690 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGTGGAGGGAGGAAATCATTAGA[A/T]AAGAAAGCAATCCCTGACCTCATGG
- Phenotype
-
MIM: 608114
MIM: 612056
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78554813] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAP1
- Gene Name
- ArfGAP with dual PH domains 1
- Gene
- GET4
- Gene Name
- golgi to ER traffic protein 4
There are no transcripts associated with this gene.
View Full Product Details