Product Details

SNP ID
rs2649903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122760312 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAATGTATATAAGGCATAGCAAA[A/G]CATTTGGCACATACTAGGTGCCCAG
Phenotype
MIM: 604550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENR PubMed Links

Gene Details

Gene
DENR
Gene Name
density regulated re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003677.4 Intron NP_003668.2

View Full Product Details