Product Details

SNP ID
rs2395084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32043712 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCCCACCCATGCCGTTTTCTTG[A/G]CTCCCACCTCTTGCCCCGGGTCCCA
Phenotype
MIM: 120820 MIM: 613815 MIM: 600985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4B PubMed Links

Gene Details

Gene
C4B
Gene Name
complement component 4B (Chido blood group)
There are no transcripts associated with this gene.

Gene
CYP21A2
Gene Name
cytochrome P450 family 21 subfamily A member 2
There are no transcripts associated with this gene.

Gene
TNXB
Gene Name
tenascin XB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019105.6 Intron NP_061978.6
NM_032470.3 Intron NP_115859.2

View Full Product Details