Product Details

SNP ID
rs4083641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:130136871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTGGTGACTCAGCCGTGCCGCA[G/T]TGGCGGGGCGGGGGACGGGGCGGCG
Phenotype
MIM: 612754
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLRX3 PubMed Links

Gene Details

Gene
GLRX3
Gene Name
glutaredoxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199868.1 Intron NP_001186797.1
NM_001321980.1 Intron NP_001308909.1
NM_006541.4 Intron NP_006532.2
Gene
LOC102724883
Gene Name
uncharacterized LOC102724883
There are no transcripts associated with this gene.

View Full Product Details