Product Details

SNP ID
rs3180499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:54459842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGTGGACCGCACGGAATTTGA[A/C]CAGTATCTGCACTTCGTGTGCAAGC
Phenotype
MIM: 610928
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100129098 PubMed Links

Gene Details

Gene
LOC100129098
Gene Name
translation initiation factor IF-2
There are no transcripts associated with this gene.

Gene
SOX17
Gene Name
SRY-box 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022454.3 1296 Missense Mutation GAA,GAC E364D NP_071899.1

View Full Product Details