Product Details

SNP ID
rs3205138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34370416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCCCACCTAAGATGGACAAAGA[G/T]GGCCACGAAAGCCAGGAGGGGGAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf24 PubMed Links

Gene Details

Gene
C9orf24
Gene Name
chromosome 9 open reading frame 24
There are no transcripts associated with this gene.

Gene
KIAA1161
Gene Name
KIAA1161
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020702.4 2695 UTR 3 NP_065753.2
XM_011517966.2 2695 UTR 3 XP_011516268.1
XM_017014930.1 2695 UTR 3 XP_016870419.1

View Full Product Details