Product Details

SNP ID
hCV27470060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101353922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTTGCTATACATCAGGACTTC[C/G]GGTGGGGACCACCGGACTGGAAATT
Phenotype
MIM: 300300 MIM: 300356
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BTK PubMed Links

Gene Details

Gene
BTK
Gene Name
Bruton tyrosine kinase
There are no transcripts associated with this gene.

Gene
TIMM8A
Gene Name
translocase of inner mitochondrial membrane 8 homolog A (yeast)
There are no transcripts associated with this gene.

View Full Product Details