Product Details

SNP ID
rs3733870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160051534 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAAAACAGTTTAGTTTCTTCATT[A/G]AATAATCATTCCTTGAGGTTCATTA
Phenotype
MIM: 601963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PWWP2A PubMed Links

Gene Details

Gene
PWWP2A
Gene Name
PWWP domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130864.1 Intron NP_001124336.1
NM_001267035.1 Intron NP_001253964.1
NM_052927.2 Intron NP_443159.1
XM_005265811.3 Intron XP_005265868.1
XM_005265812.2 Intron XP_005265869.1
XM_006714817.3 Intron XP_006714880.1
XM_011534424.2 Intron XP_011532726.1
XM_017009009.1 Intron XP_016864498.1
XM_017009010.1 Intron XP_016864499.1
XM_017009011.1 Intron XP_016864500.1
XM_017009012.1 Intron XP_016864501.1
Gene
TTC1
Gene Name
tetratricopeptide repeat domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282500.1 Intron NP_001269429.1
NM_003314.2 Intron NP_003305.1

View Full Product Details