Product Details
- SNP ID
-
rs3747423
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:85255636 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCCAAGATTGTGTTTCAGGAGTC[A/G]ACTCTCCCACAATGGTATCAGAGGA
- Phenotype
-
MIM: 314990
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF711
PubMed Links
Gene Details
- Gene
- ZNF711
- Gene Name
- zinc finger protein 711
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021998.4 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
NP_068838.3 |
XM_005262186.2 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_005262243.1 |
XM_005262187.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_005262244.1 |
XM_005262188.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_005262245.1 |
XM_005262189.2 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_005262246.1 |
XM_011531019.1 |
684 |
Missense Mutation |
AAC,GAC |
N157D |
XP_011529321.1 |
XM_011531020.1 |
684 |
Missense Mutation |
AAC,GAC |
N157D |
XP_011529322.1 |
XM_011531021.1 |
684 |
Missense Mutation |
AAC,GAC |
N157D |
XP_011529323.1 |
XM_011531022.1 |
684 |
Missense Mutation |
AAC,GAC |
N157D |
XP_011529324.1 |
XM_011531023.2 |
684 |
Missense Mutation |
AAC,GAC |
N157D |
XP_011529325.1 |
XM_011531024.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_011529326.1 |
XM_011531025.1 |
684 |
Missense Mutation |
AAC,GAC |
N157D |
XP_011529327.1 |
XM_011531026.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_011529328.1 |
XM_017029803.1 |
684 |
Missense Mutation |
AAC,GAC |
N157D |
XP_016885292.1 |
XM_017029804.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_016885293.1 |
XM_017029805.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_016885294.1 |
XM_017029806.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_016885295.1 |
XM_017029807.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_016885296.1 |
XM_017029808.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_016885297.1 |
XM_017029809.1 |
684 |
Missense Mutation |
AAC,GAC |
N153D |
XP_016885298.1 |
View Full Product Details