Product Details

SNP ID
rs3748932
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.2:100393544 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
TACGTCACCCAGTGAATGATGTGGT[C/T]CCCAAACTGAAGGTCTAGCCATCTG
Phenotype
MIM: 606376
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST10 PubMed Links

Gene Details

Gene
CHST10
Gene Name
carbohydrate sulfotransferase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004854.4 1494 Missense Mutation NP_004845.1
XM_011512207.1 1494 Missense Mutation XP_011510509.1
XM_011512208.1 1494 Missense Mutation XP_011510510.1
XM_011512210.1 1494 Missense Mutation XP_011510512.1
XM_011512211.1 1494 Missense Mutation XP_011510513.1
XM_011512212.1 1494 Missense Mutation XP_011510514.1
XM_017005380.1 1494 Missense Mutation XP_016860869.1
XM_017005381.1 1494 Missense Mutation XP_016860870.1
XM_017005382.1 1494 Missense Mutation XP_016860871.1
XM_017005383.1 1494 Missense Mutation XP_016860872.1

View Full Product Details