Product Details
- SNP ID
-
rs3751853
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31076008 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGTTGCATCTCTCTCAACTTTAA[T/C]TTCCTTAGCTATACCTGTAGGGATG
- Phenotype
-
MIM: 610561
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS53
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75489496] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRSS53
- Gene Name
- protease, serine 53
There are no transcripts associated with this gene.
- Gene
- ZNF646
- Gene Name
- zinc finger protein 646
- Gene
- ZNF668
- Gene Name
- zinc finger protein 668
There are no transcripts associated with this gene.
View Full Product Details