Product Details

SNP ID
rs3777420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159973698 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATGGTTACATTAGATCTCATGG[A/G]TTAATTCTCAGAACAGCATCCTGAC
Phenotype
MIM: 147280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGF2R PubMed Links

Gene Details

Gene
IGF2R
Gene Name
insulin like growth factor 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000876.3 Intron NP_000867.2

View Full Product Details